missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 835 98.0+%, TCI America™

Catalog No. p-7137788
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
25 mg
Catalog No. Quantity
T354025MG 25 mg
T3540100MG 100 mg
2 options

Catalog No. T354025MG

Supplier: TCI America T354025MG

Only null left

Chemical Identifiers

CAS 133550-37-5
Molecular Formula C18H16N2O3
Molecular Weight (g/mol) 308.337
MDL Number MFCD00209855
InChI Key UMGQVUWXNOJOSJ-CDCQXCROSA-N
Synonym tyrphostin b44, + enantiomer, --r-n-, a-methylbenzyl-3,4-dihydroxybenzylidenecyanoacetamide
PubChem CID 23919058
IUPAC Name (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-[(1R)-1-phenylethyl]prop-2-enamide
SMILES CC(C1=CC=CC=C1)NC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Melting Point 146°C
Quantity 25 mg
Formula Weight 308.34
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Tyrphostin AG 835
TSCA No
Recommended Storage Freezer