Learn More
Tyrphostin A9, 98+%
Selective inhibitor of PDGF receptor tyrosine kinase | CAS: 10537-47-0 | C18H22N2O | 282.387 g/mol
Supplier: Thermo Scientific Chemicals J63058MF
Description
Selective inhibitor of PDGF receptor tyrosine kinase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tyrphostin A9 | |
| Yellow | |
| MFCD00209853 | |
| UN2811 | |
| MZOPWQKISXCCTP-UHFFFAOYSA-N | |
| 2-[(3,5-ditert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile | |
| 5614 | |
| 282.39 | |
| Powder |
| 10537-47-0 | |
| C18H22N2O | |
| 50 mg | |
| tyrphostin a9, malonoben, tyrphostin 9, malonoben ansi, caswell no. 291aa, 2-3,5-di-tert-butyl-4-hydroxybenzylidene malononitrile, unii-87te8mrs65, tyrphostin 9/ag-9, 3,5-di-t-butyl-4-hydroxybenzylidene malononitrile, epa pesticide chemical code 537200 | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=C(C#N)C#N | |
| 282.387 | |
| CHEBI:82168 | |
| 99% |
Chemical Identifiers
| 10537-47-0 | |
| 282.387 | |
| MZOPWQKISXCCTP-UHFFFAOYSA-N | |
| 5614 | |
| 2-[(3,5-ditert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile |
| C18H22N2O | |
| MFCD00209853 | |
| tyrphostin a9, malonoben, tyrphostin 9, malonoben ansi, caswell no. 291aa, 2-3,5-di-tert-butyl-4-hydroxybenzylidene malononitrile, unii-87te8mrs65, tyrphostin 9/ag-9, 3,5-di-t-butyl-4-hydroxybenzylidene malononitrile, epa pesticide chemical code 537200 | |
| CHEBI:82168 | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=C(C#N)C#N |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
H301+H311-H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : OO3737000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only