missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A9, 98+%

Catalog No. AAJ63058MC
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63058MC 100 mg
AAJ63058MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63058MC Supplier Thermo Scientific Chemicals Supplier No. J63058MC

Selective inhibitor of PDGF receptor tyrosine kinase | CAS: 10537-47-0 | C18H22N2O | 282.387 g/mol

Selective inhibitor of PDGF receptor tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10537-47-0
Molecular Formula C18H22N2O
Molecular Weight (g/mol) 282.387
MDL Number MFCD00209853
InChI Key MZOPWQKISXCCTP-UHFFFAOYSA-N
Synonym tyrphostin a9, malonoben, tyrphostin 9, malonoben ansi, caswell no. 291aa, 2-3,5-di-tert-butyl-4-hydroxybenzylidene malononitrile, unii-87te8mrs65, tyrphostin 9/ag-9, 3,5-di-t-butyl-4-hydroxybenzylidene malononitrile, epa pesticide chemical code 537200
PubChem CID 5614
ChEBI CHEBI:82168
IUPAC Name 2-[(3,5-ditert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=C(C#N)C#N

Specifications

Color Yellow
Quantity 100 mg
UN Number UN2811
Formula Weight 282.39
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tyrphostin A9
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber OO3737000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only