missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A23 98.0+%, TCI America™

Catalog No. T3503100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3503100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3503100MG Supplier TCI America Supplier No. T3503100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 118409-57-7
Melting Point 222°C
Color Yellow
Molecular Formula C10H6N2O2
Quantity 100 mg
MDL Number MFCD00133899
Synonym tyrphostin 23, tyrphostin a23, 3,4-dihydroxybenzylidene malononitrile, tyrphostin ag18, tyrphostin rg50810, unii-rv0gcd31oj, 2-3,4-dihydroxyphenyl methylidene propanedinitrile, 2-3,4-dihydroxybenzylidene malononitrile, alpha-cyano-3,4-dihydroxy cinnamonitrile, propanedinitrile,2-3,4-dihydroxyphenyl methylene
InChI Key VTJXFTPMFYAJJU-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C=C(C#N)C#N)O)O
IUPAC Name 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
Molecular Weight (g/mol) 186.17
PubChem CID 2052
Formula Weight 186.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tyrphostin A23
Show More Show Less
TSCA No
Recommended Storage Refrigerator