missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A23, 99%, Thermo Scientific Chemicals

Catalog No. AAJ60308LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60308LB0 5 mg
AAJ60308MA 10 mg
AAJ60308MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60308LB0 Supplier Thermo Scientific Chemicals Supplier No. J60308LB0

Potent inhibitor of EGF receptor kinase activity

Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

Solubility
Soluble in DMSO, ethanol, or DMF

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.

Chemical Identifiers

CAS 118409-57-7
Molecular Formula C10H6N2O2
Molecular Weight (g/mol) 186.17
MDL Number MFCD00133899
InChI Key VTJXFTPMFYAJJU-UHFFFAOYSA-N
Synonym tyrphostin 23, tyrphostin a23, 3,4-dihydroxybenzylidene malononitrile, tyrphostin ag18, tyrphostin rg50810, unii-rv0gcd31oj, 2-3,4-dihydroxyphenyl methylidene propanedinitrile, 2-3,4-dihydroxybenzylidene malononitrile, alpha-cyano-3,4-dihydroxy cinnamonitrile, propanedinitrile,2-3,4-dihydroxyphenyl methylene
PubChem CID 2052
IUPAC Name 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
SMILES C1=CC(=C(C=C1C=C(C#N)C#N)O)O

Specifications

Color Brown to Yellow
Quantity 5 mg
Solubility Information Soluble in DMSO, ethanol, or DMF
Formula Weight 186.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tyrphostin A23
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only