missing translation for 'onlineSavingsMsg'
Learn More

Medisca Tyrosine (L), USP (Dietary Supplement Grade)

Catalog No. 14186222
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186222 1 kg
14185961 100 g
14186154 500 g
14185484 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14186222 Supplier Medisca Supplier No. 088309

Tyrosine (L), USP (Dietary Supplement Grade) comes as white, odorless crystals or crystalline powder. Very slightly soluble in water and insoluble in alcohol.

Tyrosine (L), USP comes as white, odorless crystals or crystalline powder. Very slightly soluble in water and insoluble in alcohol.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): l-Tyrosine

Specifications

Chemical Name or Material Tyrosine (L), USP (Dietary Supplement Grade)
Packaging Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck
CAS 60-18-4
Type Drug Standard
Molecular Weight (g/mol) 181.19
Quantity 1 kg
Percent Purity 98.5 to 101.5 %
Grade USP