missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TYROBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526861
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526861 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526861 Supplier Invitrogen™ Supplier No. PA526861
Only null left

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

DAP12 is a transmembrane adaptor protein that is non-covalently associated with several cell surface receptors on natural killer (NK), myeloid, and presumably neuronal cells. DAP12 contains an ITAM domain that preferentially recruits the protein tyrosine kinase Syk to initiate signal transduction. Deficiency of DAP12 results in reduced antigen presentation function by myeloid cells and defects in function of certain NK cell receptors in mice, as well as presenile dementia and bone cysts in humans.

Specifications

Antigen TYROBP
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TYROBP
Gene Accession No. O43914
Gene Alias DAP12; DNAX-activation protein 12; KARAP; KAR-associated protein; killer cell activating receptor associated protein; killer cell activating receptor-associated protein; killer-activating receptor-associated protein; Ly83; PLOSL; TYRO protein tyrosine kinase binding protein; TYRO protein tyrosine kinase-binding protein; Tyrobp
Gene Symbols TYROBP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 85-113 amino acids from the C-terminal region of human TYROBP.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7305
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less