missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyloxapol, USP, Spectrum™ Chemical
T1708, 25301-02-4
Supplier: Spectrum Chemical Mfg Cor T17085GM
Description
Spectrum™ Chemical Tyloxapol, USPSpecifications
| 25301-02-4 | |
| 4.0 to 7.0 | |
| Amber Glass Bottle | |
| 5 g | |
| C=O.C1CO1.CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1 | |
| 280.41 |
| 1 | |
| 0.01 | |
| C17H28O3 | |
| MDYZKJNTKZIUSK-UHFFFAOYSA-N | |
| 4-(2,4,4-trimethylpentan-2-yl)phenol; formaldehyde; oxirane | |
| USP |
Chemical Identifiers
| 25301-02-4 | |
| 280.41 | |
| 4-(2,4,4-trimethylpentan-2-yl)phenol; formaldehyde; oxirane |
| C17H28O3 | |
| MDYZKJNTKZIUSK-UHFFFAOYSA-N | |
| C=O.C1CO1.CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1 |