missing translation for 'onlineSavingsMsg'
Learn More

Tyloxapol, USP, Spectrum™ Chemical

Catalog No. p-7158427
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607619 1 g Amber Glass Bottle
18607620 5 g Amber Glass Bottle
18607621 25 g Amber Glass Bottle
18607622 100 g Amber Glass Bottle
18607623 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18607619 Supplier Spectrum Chemical Mfg Cor Supplier No. T17081GM
Only null left

T1708, 25301-02-4

Spectrum™ Chemical Tyloxapol, USP

Chemical Identifiers

CAS 25301-02-4
Molecular Formula C17H28O3
Molecular Weight (g/mol) 280.41
InChI Key MDYZKJNTKZIUSK-UHFFFAOYSA-N
IUPAC Name 4-(2,4,4-trimethylpentan-2-yl)phenol; formaldehyde; oxirane
SMILES C=O.C1CO1.CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1

Specifications

CAS Max % 1
pH 4.0 to 7.0
Ignition Residue 0.01
Packaging Amber Glass Bottle
Quantity 1 g
Grade USP