Learn More
Thermo Scientific Chemicals Tylosin tartrate
$227.48 - $227.48
Chemical Identifiers
| CAS | 74610-55-2 |
|---|---|
| Molecular Formula | C50H83NO23 |
| Molecular Weight (g/mol) | 1066.20 |
| InChI Key | ICVKYYINQHWDLM-UHFFFAOYNA-N |
| IUPAC Name | 2,3-dihydroxybutanedioic acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde |
| SMILES | OC(C(O)C(O)=O)C(O)=O.CCC1OC(=O)CC(O)C(C)C(OC2OC(C)C(OC3CC(C)(O)C(O)C(C)O3)C(C2O)N(C)C)C(CC=O)CC(C)C(=O)C=CC(C)=CC1COC1OC(C)C(O)C(OC)C1OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463070050
|
Thermo Scientific Chemicals
463070050 |
5 g |
Each for $227.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 74610-55-2 | |
| 1066.20 | |
| 2,3-dihydroxybutanedioic acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde |
| C50H83NO23 | |
| ICVKYYINQHWDLM-UHFFFAOYNA-N | |
| OC(C(O)C(O)=O)C(O)=O.CCC1OC(=O)CC(O)C(C)C(OC2OC(C)C(OC3CC(C)(O)C(O)C(C)O3)C(C2O)N(C)C)C(CC=O)CC(C)C(=O)C=CC(C)=CC1COC1OC(C)C(O)C(OC)C1OC |
Specifications
| Tylosin Tartrate | |
| 5 g | |
| Solubility aq. soln.: ca 50 g/L | |
| OC(C(O)C(O)=O)C(O)=O.CCC1OC(=O)CC(O)C(C)C(OC2OC(C)C(OC3CC(C)(O)C(O)C(C)O3)C(C2O)N(C)C)C(CC=O)CC(C)C(=O)C=CC(C)=CC1COC1OC(C)C(O)C(OC)C1OC | |
| 1066.20 | |
| Tylosin tartrate |
| 74610-55-2 | |
| C50H83NO23 | |
| ICVKYYINQHWDLM-UHFFFAOYNA-N | |
| 2,3-dihydroxybutanedioic acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde | |
| 1066.2 |
Safety and Handling
GHS H Statement May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO – Research Use Only