missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ E-3603 Tubing

Tygon™ E-3603 Tubing for laboratory and vacuum applications.

$4,701.29

Specifications

Material Tygon S3 E-3603
Overall Length (English) 50 ft.
Overall Length (Metric) 15 m
Color Clear
Product Type Tubing
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Maximum Pressure Thickness (English) Price Quantity & Availability  
14171249
Encompass
View Documents Promotion Details
Saint Gobain Life Sciences
ACF00058
0.75 in. 1.5 in. 36 psi (at 73°F) 0.375 in.
Pack of 50 Each for $4,701.29
Only null left
 
Description

Description

Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.

  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature

Compliance

FDA, NSF, 3-A, meets USP Class VI, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU.

Specifications

Specifications

Tygon S3 E-3603
15 m
Tubing
50 ft.
Clear
Videos
SDS
Documents

Documents