missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Tygon™ Clear Tubing, Formula B-44-4X DFS Item

Dense bore is smoother than bore of most stainless-steel tubing

Supplier:  Saint-Gobain AAA00038

 View more versions of this product

Catalog No. 14-169-21D


Only null left
Explore available promotions

Description

Description

  • Nontoxic, taste- and odor-free
  • Won't trap bacteria-harboring particles
  • Resistant to alkaline cleaners and unaffected by sanitizers
  • Excellent wetting properties facilitate complete drainage and permit simple flushing
  • Maximum operating temperature, 74°C (165°F)
  • Durometer hardness: Shore A, 65

Order Info

In pack of 50 ft. (15m), except as indicated in table.

Compliance

Meets FDA, 3-A Plastics Standard, and NSF Standard 51 criteria.

Specifications

Specifications

Meets FDA, 3-A Plastics Standard, and NSF Standard 51 criteria.
12.7 mm
3/4 in.
50 ft.
34 psi
1/2 in.
Tygon
19 mm
3.2 mm
165°F
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions