Learn More
Bullard™ Tychem™ QC Hoods for Respirators
For use with CC20 Airline Respirators and EVA, EVAHL, PA20, PA30, PA30IS PAPRs.
Supplier: Bullard™ 20TJ
Description
Tychem QC is a lightweight, supple material coated with polyethylene to provide excellent splash protection against many acids, bases, and other liquid chemicals. Hoods cover head, neck, and face.
Single Bib (Basic) Style: Incorporates an outer bib for overspray protection
Double Bib (Inner Bib) Style: Outer bib with a long inner bib; tuck inner bib into shirt or coveralls to funnel excess air from the hood to the body
Double Bib (Inner Bib) Style for Use with Hardhat: Accommodates Bullard™ hardhats
Specifications
| 1,000 | |
| White | |
| Respirator Hood | |
| Cc20™ series airline respirator and eva™, evahl, pa20, pa30, pa30is powered air purifying respirator | |
| 20TJ |
| NIOSH | |
| Tychem QC | |
| Pharmaceutical manufacturing, chemical and pesticide handling, painting and coating operation, nuclear operation | |
| 9 in. l bib, flex-gear™ ratchet suspension, wide angle acetate lens, headband | |
| 10/Pk. |