Learn More
DuPont™ Tychem™ 6000 SFR FS750T LT Overalls
Tychem™ 6000 SFR Bib Overall and Jacket Combo. Elastic Wrists. Open Ankles. Jacket has Double Storm Flap. Overalls have Adjustable Webbing Straps with Buckle Closure. Taped Seams. Light Tan.
Supplier: DuPont™ FS750TLT2X000400
Description
- Tychem™ 6000 SFR garments provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist.
- Designed to be worn over primary flame-resistant protective clothing, Tychem™ 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires.
- Tychem™ 6000 SFR fabric provides at least 30 min. of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
- In the event that a flash fire occurs, Tychem™ 6000 SFR garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
Specifications
| Buckle | |
| Tychem | |
| 2X-Large | |
| Open | |
| Overalls |
| Tan | |
| Taped | |
| Adjustable | |
| None | |
| Single-use |