missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 SFR FS750T LT Overalls
Tychem™ 6000 SFR Bib Overall and Jacket Combo. Elastic Wrists. Open Ankles. Jacket has Double Storm Flap. Overalls have Adjustable Webbing Straps with Buckle Closure. Taped Seams. Light Tan.
$748.63 - $908.44
Specifications
| Closure Type | Buckle |
|---|---|
| Color | Tan |
| Material | Tychem |
| Seam Type | Taped |
| Suspender | Adjustable |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-500-319
|
DuPont™
FS750TLTSM000400 |
Small |
Case of 4 Each for $748.63
|
|
|||||
|
17-500-320
|
DuPont™
FS750TLTMD000400 |
Medium |
Case of 4 Each for $748.63
|
|
|||||
|
17-500-321
|
DuPont™
FS750TLTLG000400 |
Large |
Case of 4 Each for $748.63
|
|
|||||
|
17-500-322
|
DuPont™
FS750TLTXL000400 |
X-Large |
Case of 4 Each for $748.63
|
|
|||||
|
17-500-323
|
DuPont™
FS750TLT2X000400 |
2X-Large |
Case of 4 Each for $793.55
|
|
|||||
|
17-500-324
|
DuPont™
FS750TLT3X000400 |
3X-Large |
Case of 4 Each for $841.16
|
|
|||||
|
17-500-325
|
DuPont™
FS750TLT4X000400 |
4X-Large |
Case of 4 Each for $908.44
|
|
|||||
Description
- Tychem™ 6000 SFR garments provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist.
- Designed to be worn over primary flame-resistant protective clothing, Tychem™ 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires.
- Tychem™ 6000 SFR fabric provides at least 30 min. of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
- In the event that a flash fire occurs, Tychem™ 6000 SFR garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
Specifications
| Buckle | |
| Tychem | |
| Adjustable | |
| None | |
| Single-use |
| Tan | |
| Taped | |
| Open | |
| Overalls |