Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Strong and lightweight, with broad chemical barrier
Supplier:  DuPont™ TF145TGY4X000600
 View more versions of this product
Catalog No. 19-141-754
Only null left
Explore available promotions
Alert:
Tychem™ fabrics offer little to no thermal heat protection to the user.
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3787, ASTM D2582, ASTM D5733 standards.
Specifications
Specifications
4X-Large
Tychem
Elastic
Hooded
Zipper
Standard
54.75 to 58.25 in.
139.06 to 148 cm
Use when potential exposure to chemical warfare agents exists, widely used by military first Responder™ for WMD situations
100.33 cm
6.4 to 6.7 ft.
Unisex
Gray
Elastic
Respirator Fit
Taped
Respirator fit hood, elastic wrist and ankle; Multilayer composite barrier film laminated to a Tyvek™ protective Fabric
34 in.
86.36 cm
39.5 in.
34 in.
1.95 to 2.04 m
Product Suggestions
Product Suggestions