Learn More
DuPont™ Tychem™ 5000 Coveralls with Respirator Fit Hoods, Berry Amendment Compliant
Designed for use during rigorous activities
$746.26 - $746.26
Specifications
Material Tychem
Color Tan
Disposable Single-use
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
View More Specs
Products 1
Catalog Number Mfr. No. Certifications/Compliance Closure Type Size Height (English) Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Closure Type Size Height (English) Price Quantity  
19-715-262
View Documents
DuPont™
C3128TTN2X000600
ASTM D1117 Storm Flap 2X-Large 6 to 6.4 ft.
Case of 6 Each for $746.26
Only null left
Description
Description
Coveralls protect against chemical splash as well as potential exposure to chemical warfare agents. Tan-colored polypropylene substrate offers greater physical strength and chemical holdout.
  • Multilayer barrier film laminated to a lasting 2 oz. per sq. yd.
  • Respirator-fit attached hood,
  • Front zipper closure with double storm flaps
  • Elastic wrists
  • Outer boot flaps
Compliance
Meets ASTM™ D1777 standards.
†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.
Specifications
Specifications
Tychem
Single-use
Elastic
Attached, Elastic Face
Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
0.12 kg
Tan
Attached Socks, Boot Overflaps
Hooded
Taped
0.45 mm
0.27 lb.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'