missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Coveralls with Respirator Fit Hoods, Berry Amendment Compliant

Catalog No. 19715262
Change view
Click to view available options
Certifications/Compliance:
ASTM D1117
Closure Type:
Storm Flap
Size:
2X-Large
Height (English):
6 to 6.4 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Closure Type Size Height (English)
19715262 ASTM D1117 Storm Flap 2X-Large 6 to 6.4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19715262 Supplier DuPont™ Supplier No. C3128TTN2X000600

Designed for use during rigorous activities

Coveralls protect against chemical splash as well as potential exposure to chemical warfare agents. Tan-colored polypropylene substrate offers greater physical strength and chemical holdout.

  • Multilayer barrier film laminated to a lasting 2 oz. per sq. yd.
  • Respirator-fit attached hood,
  • Front zipper closure with double storm flaps
  • Elastic wrists
  • Outer boot flaps

Compliance

Meets ASTM™ D1777 standards.

†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Specifications

Size 2X-Large
Material Tychem
Color Tan
Disposable Single-use
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Storm Flap
Seam Type Taped
Certifications/Compliance ASTM D1117
Description Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
Size (English) Chest 46.75 to 50.25 in.
Size (English) Inseam 29.75 in.
Size (Metric) Chest 118.74 to 127.63 cm
Size (Metric) Inseam 75.56 cm
Size (Metric) Waist 118.74 to 127.63 cm
Thickness 0.45 mm
For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) Sleeve 38 in.
Length (Metric) Sleeve 96.52 cm
Width (English) Chest 30 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Weight (English) 0.27 lb.
Weight (Metric) 0.12 kg
Show More Show Less