Learn More
DuPont™ Tychem™ 5000 Hooded Coverall
DuPont™ Tychem™ 5000 Coverall with Respirator Fit Hood, Attached Socks provides protection against a broad range of chemicals and situations involving liquid splash.
Supplier: DuPont™ C3128TTNMD000600
Description
- Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Attached hood with elastic around face opening.
- Strong zipper for tough, flexible closure
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| Medium | |
| Tychem | |
| Attached Socks, Boot Overflaps | |
| Zipper | |
| Respirator Fit Hood |
| Unisex | |
| Tan | |
| Elastic | |
| Taped |