missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.
Supplier: DuPont™ SL127BWHLG001200
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening.
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.
Specifications
| Large | |
| Tychem | |
| Elastic | |
| Hooded | |
| Bound | |
| 28.5 in. | |
| 12 mil | |
| 35 in. | |
| 25.75 in. | |
| 1.67 to 1.8 m | |
| 0.1 kg |
| Unisex | |
| White | |
| Elastic | |
| Zipper | |
| ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282 | |
| 97.15 to 106.04 cm | |
| Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
| 88.9 cm | |
| 5.5 to 5.9 ft. | |
| 0.22 lb. |