Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.
Supplier:  DuPont™ SL127BWH4X001200
 View more versions of this product
Catalog No. 29-635-6796
Only null left
Explore available promotions
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.
Specifications
Specifications
4X-Large
Tychem
Elastic
Hooded
Bound
32 in.
12 mil
39.5 in.
32 in.
1.95 to 2.04 m
0.1 kg
Unisex
White
Elastic
Zipper
ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
128.9 to 137.79 cm
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
100.33 cm
6.4 to 6.7 ft.
0.22 lb.
Product Suggestions
Product Suggestions