Learn More
DuPont™ Tychem™ 2000 SFR Hooded Coverall
TYCHEM™ 2000 SFR Coverall features chemical and secondary flame resistance
Supplier:  DuPont™ QS128TGRMD000400
 View more versions of this product
Catalog No. 18-888-726
Only null left
Explore available promotions
Description
Description
  • Respirator-fit hood lined with ProShield™ 6 SFR fabric.
  • Manufactured under specifications that do not contain natural rubber latex
  • Covered, braided elastic at hood.
  • Nylon zipper with large metal pull
  • Chin flap with double-sided adhesive tape
  • Storm flap covers zipper which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion at zipper.
  • Covered, braided elastic openings at wrist for tighter fit.
  • Attached socks composed of garment material.
  • Outer boot flaps with elastic are designed to cover boot tops to help reduce potential for liquid intrusion.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
Specifications
Specifications
Medium
Tychem
Single-use
Elastic
Zipper
Non-sterile
Yes
4/Pk.
Unisex
Olive Green
Attached Socks, Boot Overflaps, Elastic
Hooded
Taped
Flame, Acid
None
Product Suggestions
Product Suggestions