Learn More
DuPont™ Tychem™ 10000 Training Suit
10000 encapsulated training suits offers a high level of protection against toxic, corrosive gases and liquid chemicals
Supplier: DuPont™ TK586TLY6X000100
Description
- Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for HazMat training
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
- Extra wide (EX) single layer faceshield (20 mil PVC) for wide viewing angle
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Integrated butyl gloves
- Internal adjustable waist belt system for support and improved fit
- Clearly labeled for training use only
- Manufactured under specifications that do not contain natural rubber latex
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
| Lime Yellow | |
| 6X-Large | |
| Zipper |
| Tychem | |
| Boot Overflaps | |
| Taped |