Learn More
DuPont™ Tychem™ 10000 Training Suit
Catalog No. p-7139605
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18888881 2X-Large
18888882 3X-Large
18888883 4X-Large
18888884 5X-Large
18888885 6X-Large
18888886 Large
18888887 Medium
18888888 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18888881 Supplier DuPont™ Supplier No. TK586TLY2X000100
Only null left
10000 encapsulated training suits offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for HazMat training
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Extra wide (EX) single layer faceshield (20 mil PVC) for wide viewing angle
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Color Lime Yellow
Material Tychem
Size 2X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped