missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNL2 Monoclonal Antibody (4-1#)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566057
Description
TXNL2 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.Specifications
| TXNL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O76003, Q9CQM9, Q9JLZ1 | |
| GLRX3 | |
| Recombinant Glutaredoxin 3 (GLRX3) (Val10-Ser117). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 4-1# | |
| Unconjugated | |
| GLRX3 | |
| bA500G10.4; FLJ11864; Glrx3; GLRX4; glutaredoxin 3; glutaredoxin 4; glutaredoxin-3; GRX3; GRX4; HUSSY-22; Picot; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKCq-interacting protein; PKC-theta-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; thioredoxin-like protein 2; TXNL2; TXNL3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10539, 30926, 58815 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |