missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXK Monoclonal Antibody (K70018_2F1), Invitrogen™

Catalog No. MA556117
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556117 200 μL
MA555737 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556117 Supplier Invitrogen™ Supplier No. MA556117
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: VQKRQMRTQI SLSTDEELPE KYTQRRRPWL SQLSNKKQSN TGRVQPSKRK PLPPLPPSEV AEEKIQVKAL YDFLPREPCN LALRRAEEYL ILEKYNPHWW KARDRLGNEG LIPSNYVTEN KITNLEIYEW YHRNITRNQA EHLLRQESKE GAFIVRDSRH LGSYTISVFM GARRSTEAAI KHYQIKKNDS GQWYVAERHA FQSIPELIWY HQHNAAGLMT

TXK, along with the close relatives BMS, ITK, and BTK, signal downstream of antigen receptors and other types of receptors. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain a pleckstrin homology (PH) and a Tec homology (TH) domain in their amino termini. Four alternatively spliced forms of Tec are found to be expressed broadly in cells of hematopoietic lineage and hepatocytes. The Emt gene product associates with CD28 and becomes activated subsequent to CD28 ligation. Btk is necessary for proper B cell development, and mutations in the gene encoding Btk have been associated with families suffering from X-linked agammaglobulinemia, also referred to as Bruton's disease. The Bmx protein shares a high degree of homology with Btk and seems to be expressed at highest levels in the heart. Txk expression is T cell-specific, while expression of the Drosophila Tec homolog, Dsrc28C, is developmentally regulated.

Specifications

Antigen TXK
Applications Western Blot
Classification Monoclonal
Clone K70018_2F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TXK
Gene Accession No. P42681
Gene Alias A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK
Gene Symbols TXK
Host Species Mouse
Immunogen Recombinant human TXK (immunogen range is 21-240).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less