missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXK Monoclonal Antibody (K70018_2F1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555737
Description
Sequence of this protein is as follows: VQKRQMRTQI SLSTDEELPE KYTQRRRPWL SQLSNKKQSN TGRVQPSKRK PLPPLPPSEV AEEKIQVKAL YDFLPREPCN LALRRAEEYL ILEKYNPHWW KARDRLGNEG LIPSNYVTEN KITNLEIYEW YHRNITRNQA EHLLRQESKE GAFIVRDSRH LGSYTISVFM GARRSTEAAI KHYQIKKNDS GQWYVAERHA FQSIPELIWY HQHNAAGLMT
TXK, along with the close relatives BMS, ITK, and BTK, signal downstream of antigen receptors and other types of receptors. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain a pleckstrin homology (PH) and a Tec homology (TH) domain in their amino termini. Four alternatively spliced forms of Tec are found to be expressed broadly in cells of hematopoietic lineage and hepatocytes. The Emt gene product associates with CD28 and becomes activated subsequent to CD28 ligation. Btk is necessary for proper B cell development, and mutations in the gene encoding Btk have been associated with families suffering from X-linked agammaglobulinemia, also referred to as Bruton's disease. The Bmx protein shares a high degree of homology with Btk and seems to be expressed at highest levels in the heart. Txk expression is T cell-specific, while expression of the Drosophila Tec homolog, Dsrc28C, is developmentally regulated.Chemical Identifiers
| TXK | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK | |
| Recombinant human TXK (immunogen range is 21-240). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TXK |
| K70018_2F1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P42681 | |
| 7294 | |
| IgG2a | |
| Protein A | |
| Antibody |
Specifications
| TXK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P42681 | |
| TXK | |
| Recombinant human TXK (immunogen range is 21-240). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K70018_2F1 | |
| Unconjugated | |
| TXK | |
| A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK | |
| Mouse | |
| Protein A | |
| RUO | |
| 7294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |