Learn More
Thermo Scientific™ TX-750 4 x 750 mL Swinging Bucket Rotor
Increase lab productivity with the 750 mL swinging bucket rotor, delivering maximum flexibility at 4700 rpm.
$388.79 - $8,489.13
Specifications
Certifications/Compliance Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Biocontainment Yes
View More Specs
Products 8
Catalog Number Mfr. No. Capacity Type For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Type For Use With (Equipment) Price Quantity & Availability  
75003180
View Documents Promotion Details
Thermo Scientific™
75003180
4 x 750 mL Swinging Bucket Rotor Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Each for $7,169.38
Only null left
75003608
View Documents Promotion Details
Thermo Scientific™
75003608
750 mL Swinging Bucket Rotor Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Pack of 4 Each for $5,920.98
Only null left
75003609
View Documents Promotion Details
Thermo Scientific™
75003609
Not Applicable Bucket Lid Round Buckets
Pack of 4 Each for $1,690.42
Only null left
75003610
View Documents Promotion Details
Thermo Scientific™
75003610
Not Applicable O-ring ClickSeal Biocontainment Lids
Each for $388.79
Only null left
75003614
View Documents Promotion Details
Thermo Scientific™
75003614
250 mL Swinging Bucket Rotor Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Pack of 4 Each for $8,489.13
Only null left
75003615
View Documents Promotion Details
Thermo Scientific™
75003615
Not Applicable Bucket Lid Rectangular Buckets
Each for $1,592.60
Only null left
75003617
View Documents Promotion Details
Thermo Scientific™
75003617
4 x 750 mL Microplate/Flask Carrier Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Each for $6,830.54
Only null left
75003795
View Documents Promotion Details
Thermo Scientific™
75003795
4 x 750 mL Microplate/Flask Carrier Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Each for $3,317.17
Only null left
Description
Description
Increase lab productivity with the Thermo Scientific™ TX-750 4 x 750mL Swinging Bucket Rotor, delivering maximum flexibility at 4700rpm.
Ordering Information:
Buckets, lids and microplate carriers are sold separately; order TX-750 rotor body (75003180) and components to complete assembly. For replacement O-rings for lids (set of four) order 75003610. Tube adapters are also available separately. Contact your sales representative for information.
Includes:
Rotor manual
Compatible with:
Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Specifications
Specifications
Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Yes