Learn More
Thermo Scientific™ TX-750 4 x 750 mL Swinging Bucket Rotor
Increase lab productivity with the 750 mL swinging bucket rotor, delivering maximum flexibility at 4700 rpm.
Supplier: Thermo Scientific™ 75003617
Description
Increase lab productivity with the Thermo Scientific™ TX-750 4 x 750mL Swinging Bucket Rotor, delivering maximum flexibility at 4700rpm.
Ordering Information:Buckets, lids and microplate carriers are sold separately; order Rotor Body (75-003-607) and components to complete assembly. For replacement O-rings for lids (set of four) order 75-003-610. Tube adapters are also available separately. Contact your sales representative for information.
Includes:
Rotor manual
Compatible with:
Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Chemical Identifiers
| Biocontainment certified by CAMR™, HPA, Porton Down, U.K. | |
| Yes |
Specifications
| Rubber (Pad) | |
| 4 x 750 mL | |
| Biocontainment certified by CAMR™, HPA, Porton Down, U.K. | |
| Set of 4 |
| Microplate/Flask Carrier | |
| Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F | |
| Yes |