Learn More
Thermo Scientific™ TX-400 Rotor Adapters
Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.
$1,280.50 - $1,396.02
Specifications
No. per Pack 4
View More Specs
Products 15
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
75003680
View Documents Promotion Details
Thermo Scientific™
75003680
76 x 5/7 mL Blood Collection Tube
Each for $1,325.75
Only null left
75003681
View Documents Promotion Details
Thermo Scientific™
75003681
56 x 10/15 mL Blood Collection Tube
Each for $1,350.44
Only null left
75003682
View Documents Promotion Details
Thermo Scientific™
75003682
15 mL Conical Tube
Each for $1,363.73
Only null left
75003683
View Documents Promotion Details
Thermo Scientific™
75003683
50 mL Conical Tube
Each for $1,350.44
Only null left
75003700
View Documents Promotion Details
Thermo Scientific™
75003700
1.5/2.0 mL Conical Microtube
Each for $1,344.75
Only null left
75003703
View Documents Promotion Details
Thermo Scientific™
75003703
30/25 mL DIN Round/Flat-bottom
Each for $1,396.02
Only null left
75003704
View Documents Promotion Details
Thermo Scientific™
75003704
15 mL Round-bottom Sarstedt
Each for $1,344.75
Only null left
75003706
View Documents Promotion Details
Thermo Scientific™
75003706
25 mL Universal
Each for $1,280.50
Only null left
75003707
View Documents Promotion Details
Thermo Scientific™
75003707
50 mL DIN Round-bottom
Each for $1,363.73
Only null left
75003708
View Documents Promotion Details
Thermo Scientific™
75003708
100 mL Round-bottom, Open-tube
Each for $1,369.44
Only null left
75003788
View Documents Promotion Details
Thermo Scientific™
75003788
175, 200, 225, 250 mL Bottle
Each for $1,363.73
Only null left
75003793
View Documents Promotion Details
Thermo Scientific™
75003793
3 mL RIA or Round-bottom without Cap
Each for $1,320.06
Only null left
75003794
View Documents Promotion Details
Thermo Scientific™
75003794
15 mL Blood Collection Tube
Each for $1,308.65
Only null left
75003798
View Documents Promotion Details
Thermo Scientific™
75003798
14 mL Conical Urine Tube
Each for $1,344.75
Only null left
75003799
View Documents Promotion Details
Thermo Scientific™
75003799
50 mL Round-bottom
Each for $1,310.56
Only null left
Description
Description
Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 × 250mL (4 × 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. × L: 62 × 125mm (80 × 125mm with 400mL bio-bottles)
Specifications
Specifications
4