missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TX-400 Rotor Adapters

Catalog No. 75003680
Change view
Click to view available options
Capacity:
1.5/2.0 mL Conical Microtube
100 mL Round-bottom, Open-tube
14 mL Conical Urine Tube
15 mL Blood Collection Tube
15 mL Conical Tube
15 mL Round-bottom Sarstedt
175, 200, 225, 250 mL Bottle
3 mL RIA or Round-bottom without Cap
30/25 mL DIN Round/Flat-bottom
50 mL Conical Tube
50 mL DIN Round-bottom
50 mL Round-bottom
56 x 10/15 mL Blood Collection Tube
76 x 5/7 mL Blood Collection Tube
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75003680 76 x 5/7 mL Blood Collection Tube
75003681 56 x 10/15 mL Blood Collection Tube
75003682 15 mL Conical Tube
75003683 50 mL Conical Tube
75003700 1.5/2.0 mL Conical Microtube
75003703 30/25 mL DIN Round/Flat-bottom
75003704 15 mL Round-bottom Sarstedt
75003707 50 mL DIN Round-bottom
75003708 100 mL Round-bottom, Open-tube
75003788 175, 200, 225, 250 mL Bottle
75003793 3 mL RIA or Round-bottom without Cap
75003794 15 mL Blood Collection Tube
75003798 14 mL Conical Urine Tube
75003799 50 mL Round-bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 75003680 Supplier Thermo Scientific™ Supplier No. 75003680

Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.

Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.

The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 × 250mL (4 × 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. × L: 62 × 125mm (80 × 125mm with 400mL bio-bottles)

Specifications

Type Round Bucket Adapter
Tube Type 5/7 mL Blood Collection Tube
Capacity 76 x 5/7 mL Blood Collection Tube
Max. Tube Diameter 13 mm
Max. Tube Length 110 mm
For Use With (Equipment) TX-400 Rotor
No. per Pack 4