Learn More
Thermo Scientific™ TX-200 Swinging Bucket Rotor
Accommodate a wide range of processing volumes with the TX-200 4 x 180mL Swinging Bucket Rotor.
$410.19 - $3,121.00
Specifications
Biocontainment Yes
View More Specs
Products 4
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
75003658
Accessories View Documents Promotion Details
Thermo Scientific™
75003658
Swinging Bucket Rotor
Each for $3,121.00
Only null left
75003659
Accessories View Documents Promotion Details
Thermo Scientific™
75003659
Bucket
Each for $2,015.27
Only null left
75003660
Accessories View Documents Promotion Details
Thermo Scientific™
75003660
Bucket Lid
Each for $550.81
Only null left
75003687
View Documents Promotion Details
Thermo Scientific™
75003687
Replacement O-ring
Pack of 4 Each for $410.19
Only null left
Description
Description
Accommodate a wide range of processing volumes with the Thermo Scientific™ TX-200 4 x 180mL Swinging Bucket Rotor.
Ordering Information:
Round buckets (75-003-659), biocontainment lids (75-003-660) and replacement O-rings (75-003-687) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Specifications
Specifications
Yes