Learn More
Fisherbrand™ Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
$190.96 - $199.61
Specifications
| Material | Natural Rubber |
|---|---|
| Product Type | Twistit Stopper |
| Stopper Number | Assorted |
| For Use With (Application) | Multi-purpose applications |
| Size | Assorted |
Description
- Simple twist-off seals provide convenient one, two, or three-hole use
- Solid stoppers are vacuum-tested at 30 in. of mercury
- High resistance to deformation—even after repeated autoclaving
- Each stopper has its size number in raised figures on top
Specifications
| Natural Rubber | |
| Assorted | |
| Assorted |
| Twistit Stopper | |
| Multi-purpose applications |