missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Twistit™ Rubber Stoppers

Catalog No. 14131J
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14131J Black
14132J White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14131J

Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation

  • Simple twist-off seals provide convenient one, two, or three-hole use
  • Solid stoppers are vacuum-tested at 30 in. of mercury
  • High resistance to deformation—even after repeated autoclaving
  • Each stopper has its size number in raised figures on top

Specifications

Product Type Twistit Stopper
Stopper Number Assorted
Description Stopper No. 3 (9 Ea.), No. 4 (8 Ea.), No. 5 (6 Ea.), No. 6 (4 Ea.), No. 7 (2 Ea.), No. 8 (2 Ea.), No. 9 (1 Ea.), No. 10 (1 Ea.)
For Use With (Application) Multi-purpose applications
Size Assorted
Color Black
Material Natural Rubber