missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 85, MP Biomedicals
$185.90
Chemical Identifiers
| CAS | 87-44-5 |
|---|---|
| Molecular Formula | C15H24 |
| Molecular Weight (g/mol) | 204.357 |
| InChI Key | NPNUFJAVOOONJE-IZZDOVSWSA-N |
| Synonym | Polyoxyethylenesorbitan Trioleate |
| PubChem CID | 5354499 |
| IUPAC Name | (4E)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene |
| SMILES | CC1=CCCC(=C)C2CC(C2CC1)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN15248790
|
N/A | 500 mL |
Each for $185.90
|
|
|||||
Chemical Identifiers
| 87-44-5 | |
| 204.357 | |
| Polyoxyethylenesorbitan Trioleate | |
| (4E)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene |
| C15H24 | |
| NPNUFJAVOOONJE-IZZDOVSWSA-N | |
| 5354499 | |
| CC1=CCCC(=C)C2CC(C2CC1)(C)C |
Specifications
| 87-44-5 | |
| Polyoxyethylenesorbitan Trioleate | |
| CC1=CCCC(=C)C2CC(C2CC1)(C)C | |
| 204.357 | |
| ≈1840 |
| C15H24 | |
| NPNUFJAVOOONJE-IZZDOVSWSA-N | |
| (4E)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene | |
| 5354499 | |
| Tween 85 |
Safety and Handling
Recommended Storage : Store at room temperature.