missing translation for 'onlineSavingsMsg'
Learn More

Tween 85, MP Biomedicals

Catalog No. ICN15248790
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15248790 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15248790 Supplier MP Biomedicals Inc Supplier No. 0215248790

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-44-5
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
InChI Key NPNUFJAVOOONJE-IZZDOVSWSA-N
Synonym Polyoxyethylenesorbitan Trioleate
PubChem CID 5354499
IUPAC Name (4E)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene
SMILES CC1=CCCC(=C)C2CC(C2CC1)(C)C

Specifications

Quantity 500 mL
Formula Weight ≈1840
Chemical Name or Material Tween 85
Recommended Storage Store at room temperature.