missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 80 (=Polyoxyethylene Sorbitan Monooleate) [for Biochemical Research], TCI America™
Supplier: TCI America T253325G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 9005-65-6 | |
| 604.82 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| MFCD00082107 | |
| Polyoxyethylene Sorbitan Monooleate | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Specifications
| Tween 80 (=Polyoxyethylene Sorbitan Monooleate) [for Biochemical Research] | |
| 9005-65-6 | |
| -25°C | |
| 25 g | |
| MFCD00082107 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
| Yellow | |
| Clear Liquid at 20°C | |
| 5 to 7 (5% H2O soln.) | |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| Polyoxyethylene Sorbitan Monooleate | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 604.82 |
Safety and Handling
EINECSNumber : (8)-0055
RTECSNumber : WG2932500
TSCA : Yes