missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Tween™ 80, Fisher BioReagents
Non-ionic detergent for selective protein extraction
Manufacturer: Fisher BioReagents BP338500
Description
This nonionic detergent is used in the isolation of water-soluble membrane proteins.Specifications
Tween™ 80 | |
Vapor Pressure: <1hPa at 20°C | |
0.0005% max. | |
65 to 80 | |
0.25% max. | |
Polyoxyethylene-20-sorbitan Monooleate, Polysorbate 80 | |
HDPE Bottle | |
3% max. | |
6 |
9005-65-6 | |
0.0001% max. | |
1.080g/cm³ | |
Pass Test | |
15, 7703 | |
0.0005% max. | |
Liquid | |
Amber | |
500mL |
Chemical Identifiers
9005-65-6 | |
Polyoxyethylene-20-sorbitan Monooleate, Polysorbate 80 |
Safety and Handling

Recommended Storage : RT
Promotions
-
Fisher BioReagents
Get 2 Extra Life Sciences Chemicals When You Buy 3
Buy three life sciences chemicals and get two more at no additional cost.: 03/31/21