missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween™80, ∽75% oleic acid, MP Biomedicals™
Chemical Identifiers
| CAS | 9005-70-3 |
|---|---|
| Molecular Formula | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 |
| Molecular Weight (g/mol) | NaN |
| MDL Number | MFCD01779641 |
| InChI Key | ZGEQUSWSYZZNAI-CLFAGFIQNA-N |
| Synonym | Polysorbate 80, Polyoxyethylene sorbitan monooleate |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10317090
|
N/A | 0.5 L |
N/A
|
|
|||||
|
ICN19472504
|
N/A | 4 L |
N/A
|
|
|||||
Chemical Identifiers
| 9005-70-3 | |
| NaN | |
| ZGEQUSWSYZZNAI-CLFAGFIQNA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 | |
| MFCD01779641 | |
| Polysorbate 80, Polyoxyethylene sorbitan monooleate |
Specifications
| Yellow | |
| Liquid | |
| ∽75% | |
| MFCD01779641 | |
| ZGEQUSWSYZZNAI-CLFAGFIQNA-N | |
| NaN |
| 9005-70-3 | |
| 1.08g/mL | |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 | |
| Polysorbate 80, Polyoxyethylene sorbitan monooleate | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO | |
| Tween 80 |
Safety and Handling
Recommended Storage : Store at room temperature.