missing translation for 'onlineSavingsMsg'
Learn More

Tween™80, ∽75% oleic acid, MP Biomedicals™

Catalog No. ICN19472504
Change view
Click to view available options
Quantity:
0.5 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19472504 4 L
ICN10317090 0.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19472504 Supplier MP Biomedicals Inc Supplier No. 0219472504

Chemical Identifiers

CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polysorbate 80, Polyoxyethylene sorbitan monooleate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Liquid
Density 1.08g/mL
Quantity 4 L
Assay Percent Range ∽75%
Chemical Name or Material Tween 80
Recommended Storage Store at room temperature.