missing translation for 'onlineSavingsMsg'
Learn More

Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate), TCI America™

$39.03 - $66.36

Chemical Identifiers

CAS 9005-66-7
Molecular Formula (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165345
InChI Key DVRFSZFBSOKHQW-UHFFFAOYNA-N
Synonym Polyoxyethylene Sorbitan Monopalmitate
PubChem CID 92329579
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
T054425G
View Documents Promotion Details
N/A 25 g
Each for $39.03
Only null left
 
T0544500G
View Documents Promotion Details
N/A 500 g
Each for $66.36
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9005-66-7
522.68
DVRFSZFBSOKHQW-UHFFFAOYNA-N
92329579
CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
MFCD00165345
Polyoxyethylene Sorbitan Monopalmitate
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate

Specifications

Yellow
Clear Liquid-Cloudy Liquid at 20°C
MFCD00165345
DVRFSZFBSOKHQW-UHFFFAOYNA-N
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
92329579
9005-66-7
(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Polyoxyethylene Sorbitan Monopalmitate
CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0055

RTECSNumber : WG2933000

TSCA : Yes

SDS
Documents

Documents