missing translation for 'onlineSavingsMsg'
Learn More

Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate), TCI America™

Catalog No. T054425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T054425G 25 g
T0544500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T054425G Supplier TCI America Supplier No. T054425G

Chemical Identifiers

CAS 9005-66-7
Molecular Formula (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165345
InChI Key DVRFSZFBSOKHQW-UHFFFAOYNA-N
Synonym Polyoxyethylene Sorbitan Monopalmitate
PubChem CID 92329579
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Clear Liquid-Cloudy Liquid at 20°C
Quantity 25 g
Chemical Name or Material Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate)
EINECSNumber (8)-0055
RTECSNumber WG2933000
TSCA Yes