missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate) [for Biochemical Research], TCI America™
$33.18 - $64.41
Chemical Identifiers
| CAS | 9005-66-7 |
|---|---|
| Molecular Formula | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165345 |
| InChI Key | DVRFSZFBSOKHQW-UHFFFAOYNA-N |
| Synonym | Polyoxyethylene Sorbitan Monopalmitate |
| PubChem CID | 92329579 |
| IUPAC Name | 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Chemical Identifiers
| 9005-66-7 | |
| 522.68 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| 92329579 | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| MFCD00165345 | |
| Polyoxyethylene Sorbitan Monopalmitate | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Specifications
| Yellow | |
| Clear Liquid-Cloudy Liquid at 20°C | |
| (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | |
| Polyoxyethylene Sorbitan Monopalmitate | |
| CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate) [for Biochemical Research] |
| 9005-66-7 | |
| 20°C | |
| MFCD00165345 | |
| DVRFSZFBSOKHQW-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
| 92329579 |
Safety and Handling
EINECSNumber : (8)-0055
RTECSNumber : WG2933000
TSCA : Yes