missing translation for 'onlineSavingsMsg'
Learn More

Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate) [for Biochemical Research], TCI America™

Supplier:  TCI America T253125G

Quantity 25 g

 View more versions of this product

Catalog No. T253125G

Only null left

Description

Description

Specifications

Chemical Identifiers

9005-66-7
522.68
DVRFSZFBSOKHQW-UHFFFAOYNA-N
92329579
CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
MFCD00165345
Polyoxyethylene Sorbitan Monopalmitate
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate

Specifications

Tween 40 (=Polyoxyethylene Sorbitan Monopalmitate) [for Biochemical Research]
9005-66-7
20°C
(C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Polyoxyethylene Sorbitan Monopalmitate
CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Yellow
Clear Liquid-Cloudy Liquid at 20°C
25 g
MFCD00165345
DVRFSZFBSOKHQW-UHFFFAOYNA-N
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
92329579
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0055

RTECSNumber : WG2933000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions