missing translation for 'onlineSavingsMsg'
Learn More

Tween 20, Molecular Biology Grade, Promega™

Catalog No. PRH5152
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRH5152 100 mL
PRH5151 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRH5152 Supplier Promega Corporation Supplier No. H5152

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A nonionic detergent used in many molecular biology applications.

  • Quality tested and certified free of DNase, RNase and protease activity

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym tween 20, polysorbate 20, polyoxyethylene sorbitan monolaurate, polyoxyethylene 20 sorbitan monolaurate, polysorbate inn, 2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate, alkest tw 20, polysorbate 20 nf, polysorbate 40 nf
PubChem CID 443314
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow to Amber
Density 1.083g/cu.cm
Quantity 100 mL
Physical Form Viscous Liquid
Formula Weight 1163.9
Chemical Name or Material Tween 20
Recommended Storage 15 to 30°C