missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TurboFect™ Transfection Reagent

Catalog No. ferr0533
Change view
Click to view available options
Quantity:
200 μL
1 mL
5 x 200 μL
5 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0533 200 μL
FERR0534 5 x 200 μL
FERR0531 1 mL
FERR0532 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERR0533 Supplier Thermo Scientific™ Supplier No. R0533
Only null left

Deliver DNA plasmids and expression vectors into eukaryotic cells with this highly efficient, easy-to-use, non-immunogenic transfection reagent.

The Thermo Scientific™ TurboFect in vitro Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.

TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery into eukaryotic cells. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells. Transfection can be performed in the presence or absence of serum. TurboFect Reagent demonstrates superior transfection efficiency and minimal toxicity when compared to other lipid-based or polymer-based transfection reagents.

Highlights:

  • High transfection efficiency of a wide variety of cell types
  • No optimization required for most cell lines
  • Excellent transfection efficiency in the presence or absence of serum
  • Minimal cytotoxicity
  • Ready-to-use – no need to reconstitute, dilute or manipulate

Recommended for:

Stable and transient transfection or co-transfection of DNA or RNA; Experiments with adherent or suspension cells, differentiated or undifferentiated cells; Gene expression studies and assays involving GFP or luciferase reporters

Specifications

Type Transfection Reagent
Sufficient For 100 Transfections
Quantity 200 μL
Content And Storage Upon receipt store at 4°C.
Transfection Technique Lipid-Based Transfection
Product Line TurboFect

For Research Use Only. Not for use in diagnostic procedures.