missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TurboFect™ Transfection Reagent
Description
The Thermo Scientific™ TurboFect in vitro Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery into eukaryotic cells. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells. Transfection can be performed in the presence or absence of serum. TurboFect Reagent demonstrates superior transfection efficiency and minimal toxicity when compared to other lipid-based or polymer-based transfection reagents.
Highlights:
- High transfection efficiency of a wide variety of cell types
- No optimization required for most cell lines
- Excellent transfection efficiency in the presence or absence of serum
- Minimal cytotoxicity
- Ready-to-use – no need to reconstitute, dilute or manipulate
Recommended for:
Stable and transient transfection or co-transfection of DNA or RNA; Experiments with adherent or suspension cells, differentiated or undifferentiated cells; Gene expression studies and assays involving GFP or luciferase reporters
Specifications
Specifications
| Type | Transfection Reagent |
| Sufficient For | 250 Transfections |
| Quantity | 5 x 1 mL |
| Content And Storage | Upon receipt store at 4°C. |
| Classification | Chemically Defined |
| Compatible Cells | Established Cell Lines, Primary Cells, Endothelial Cells, Epithelial Cells, Kidney, Macrophages, Myeloma Cells, Hepatocytes, Fibroblasts |
| Starting Material | Plasmid DNA |
| Transfection Technique | Chemical Transfection, Lipid-Based Transfection |
| Product Line | TurboFect |
For Research Use Only. Not for use in diagnostic procedures.