missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TurboFect™ Transfection Reagent
Description
The Thermo Scientific™ TurboFect in vitro Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery in vivo. Reagent causes no detectable inflamatory response and is suitable for DNA administration via various routes, including intravenous, intraperitoneal, intratumoral injection, etc.
- No detectable inflamatory response
- Gene delivery via multiple routes of administration
- Reproducible results
- Functionally tested in mice and rats
Recommended for:
DNA transfection
siRNA-DNA co-transfection
Specifications
Specifications
| Type | Transfection Reagent |
| Quantity | 1 mL |
| Classification | Chemically Defined |
| Compatible Cells | Primary Cell, Endothelial Cell, Epithelial Cell, Myeloma Cell, Cell Lines |
| Format | 6-Well Plate, 12-Well Plate, 24-Well Plate, 48-Well Plate, 96-Well Plate, Flasks |
| Transfection Technique | Chemical Transfection |
| Product Line | TurboFect |
| For Use With (Application) | Transfection |
For Research Use Only. Not for use in diagnostic procedures.