missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Stock Solution II, Hexamethylene Tetramine, 10%, LabChem™

Catalog No. p-5388073
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC263207 125 mL
LC263201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC263207 Supplier LabChem Supplier No. LC263207
Only null left

Specifications

Chemical Name or Material Turbidity Stock Solution II
Name Note Hexamethylene Tetramine
CAS 100-97-0, 7732-18-5
Color Colorless
CAS Max % 90, 10
Decomposition Information Gaseous Ammonia; Nitrogen oxides
Physical Form Liquid
Quantity 125 mL
Identification Passes Test
Linear Formula C6H12N4
Packaging Poly Bottle
Solubility Information Soluble in water
Formula Weight 140.19
Percent Purity 10%
Show More Show Less