missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Standard, 0 NTU (< 0.1 Nephelometric Turbidity Units), Ricca Chemical

Catalog No. p-7239416
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8804000500 500 mL
R88040001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R8804000500 Supplier Ricca Chemical Company Supplier No. R8804000500C
Only null left

Specifications

Chemical Name or Material Turbidity Standard
Packaging Amber Glass Bottle
Name Note 0.1 NTU
CAS 26628-22-8, 7732-18-5
Odor Odorless
Assay Percent Range Turbidity: < 0.1 NTU
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solubility Infinitely soluble in water
Solvent or Matrix Water
Quantity 500 mL
Show More Show Less